Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST traceable, Orion™

pH buffer solutions, NIST traceable, Orion™

pH 7,00 buffer, colour-coded yellow (pH Buffer Cubitainers™), 7,00, Yellow, Plastic bottle, 19 L
Catalog # ORIO9107CB
Supplier:   Thermo Orion
pH buffer solutions, NIST traceable, Orion™
Catalog # ORIO9107CB
Supplier:  Thermo Orion
CAS Number:  
pH buffer solutions, NIST traceable, Orion™
pH buffer solutions, NIST traceable, Orion™

pH 7,00 buffer, colour-coded yellow (pH Buffer Cubitainers™), 7,00, Yellow, Plastic bottle, 19 L

CAS Number:  
Price
211,00 €
Log in for availability
1 * 19 L
Product Details & Documents

Perform accurate, reproducible pH calibrations with Orion™ pH buffers.

  • NIST traceability
  • Colour-coded solutions are available for ease of use and identification

Solutions are manufactured under ISO 9000 quality standards.

The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.

Delivery information: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    19 L
  • Colour:
    Yellow
  • Description:
    pH 7,00 buffer, colour-coded yellow (pH Buffer Cubitainers™)
  • pH value:
    7,00

Specifications