Magnetic stirring bars, triangular
33,83, 8,13 mm
Catalog # KART782
Supplier: KARTELL
Jump to:
Product Details & Documents
PTFE coated, triangular magnetic stirring bars.
- Scraping action ideal for dissolving solids and mixing sediments
- Angled surfaces create high turbulence even at low speeds
- Suitable for food stuff
Specifications
- Ø:8,13 mm
- Length:33,83
- Seq:03