Orders LinkContactUsLinkComponent
Filter candles
Filter candles
4, 13×25 mm
Catalog # LENZ07206425
Supplier:   Lenz Laborglas
Filter candles
Catalog # LENZ07206425
Supplier:  Lenz Laborglas
Filter candles
Filter candles
4, 13×25 mm
Price
43,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
Specifications
  • Ø×H:
    13×25 mm
  • Porosity:
    4
  • Seq:
    10