Orders LinkContactUsLinkComponent
Filter candles
Filter candles
1, 9×20 mm
Catalog # LENZ07205120
Supplier:   Lenz Laborglas
Filter candles
Catalog # LENZ07205120
Supplier:  Lenz Laborglas
Filter candles
Filter candles
1, 9×20 mm
Price
32,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
Specifications
  • Ø×H:
    9×20 mm
  • Porosity:
    1
  • Seq:
    02