Flasks, flat bottom, wide neck, with beaded rim
2000 ml, 230 mm, 50 mm, 166 mm
Catalog # LENZ07050073
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck external diameter larger than 65 mm have reinforced rims.
Certifications: Acc. to DIN ISO 24450
Documents
BSE TSE Glass Columns
Certificats BSE/TSE
Specifications
- Ext.Ø:166 mm
- Capacity:2000 ml
- Height:230 mm
- Seq:06
- Neck Ø ext.:50 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



