Orders LinkContactUsLinkComponent
O-Ring seals for flat flanges
O-Ring seals for flat flanges
Flat flanges DN 200
Catalog # LENZ06248220
Supplier:   Lenz Laborglas
O-Ring seals for flat flanges
Catalog # LENZ06248220
Supplier:  Lenz Laborglas
O-Ring seals for flat flanges
O-Ring seals for flat flanges
Flat flanges DN 200
Price
77,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Silicone or Silicone PTFE-coated.
  • Excellent chemical resistance
  • Excellent temperature resistance
Seals for flat flanges LF 60-200.
Specifications
  • For:
    Flat flanges DN 200
  • Seq:
    08