Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Cartouches d'extraction
Cartouches d'extraction
Extraction insert for solids, for extractor 250 ml capacity (500 ml volume)
Catalog # LENZ05431203
Supplier:   Lenz Laborglas
Cartouches d'extraction
Catalog # LENZ05431203
Supplier:  Lenz Laborglas
Cartouches d'extraction
Cartouches d'extraction
Extraction insert for solids, for extractor 250 ml capacity (500 ml volume)
Price
54,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Glass insert with bores for solids.
Documents
Specifications
  • Désignation:
    Extraction insert for solids, for extractor 250 ml capacity (500 ml volume)
  • Seq:
    12