Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
Extraction insert for solids, for extractor 70 ml capacity (100 ml volume)
Catalog # LENZ05431201
Supplier:   Lenz Laborglas
Extraction thimbles
Catalog # LENZ05431201
Supplier:  Lenz Laborglas
Extraction thimbles
Extraction thimbles
Extraction insert for solids, for extractor 70 ml capacity (100 ml volume)
Price
42,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Glass insert with bores for solids.
Documents
Specifications
  • Description:
    Extraction insert for solids, for extractor 70 ml capacity (100 ml volume)
  • Seq:
    10