Order Entry
Belgium
Orders LinkContactUsLinkComponent
Extractors for solids

Extractors for solids

100 ml, 250 ml, 45/40 NS
Catalog # LENZ05431037
Supplier:   Lenz Laborglas
Extractors for solids
Catalog # LENZ05431037
Supplier:  Lenz Laborglas
Extractors for solids
Extractors for solids

100 ml, 250 ml, 45/40 NS

Price
268,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass.

  • Very good chemical resistance
  • Very good temperature resistance

For the extraction of solids (warm).

The complete apparatuses are consisting of:
Extractor head with glass thimbles for solids, Dimroth condenser, with thread GL 14, round bottom flask.

Documents

Specifications
  • Extraction cap.:
    100 ml
  • Socket size:
    45/40 NS
  • Seq:
    02
  • Flask capacity:
    250 ml