Order Entry
Belgium
Orders LinkContactUsLinkComponent
Extractors for solids

Extractors for solids

70 ml, 250 ml, 45/40 NS
Catalog # LENZ05431031
Supplier:   Lenz Laborglas
Extractors for solids
Catalog # LENZ05431031
Supplier:  Lenz Laborglas
Extractors for solids
Extractors for solids

70 ml, 250 ml, 45/40 NS

Price
262,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass.

  • Very good chemical resistance
  • Very good temperature resistance

For the extraction of solids (warm).

The complete apparatuses are consisting of:
Extractor head with glass thimbles for solids, Dimroth condenser, with thread GL 14, round bottom flask.

Documents

Specifications
  • Extraction cap.:
    70 ml
  • Socket size:
    45/40 NS
  • Seq:
    01
  • Flask capacity:
    250 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.