Orders LinkContactUsLinkComponent
Bomb sampler, Sub Sampler Pro
Bomb sampler, Sub Sampler Pro
Bomb samplers, Nickel plated brass, 500 ml
Catalog # SAMP5072A-500
Supplier:   Sampling Systems
Bomb sampler, Sub Sampler Pro
Catalog # SAMP5072A-500
Supplier:  Sampling Systems
Bomb sampler, Sub Sampler Pro
Bomb sampler, Sub Sampler Pro
Bomb samplers, Nickel plated brass, 500 ml
Price
1 210,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Essential for flammable liquids as a heavy-duty sampler.
  • Metallic parts manufactured from stainless steel
  • Unique double action to sample from a set distance
  • Can be plunged until the assembly touches the bottom of the container
Specially designed for the environment throughout the world for both chemical and environmental sampling.
Certifications: Conforms to FDA CFR 177.2600.
Specifications
  • Capacity:
    500 ml
  • Description:
    Bomb samplers
  • Material:
    Nickel plated brass