Orders LinkContactUsLinkComponent
Embedding rings
Embedding rings
White
Catalog # KART2919
Supplier:   KARTELL
Embedding rings
Catalog # KART2919
Supplier:  KARTELL
Embedding rings
Embedding rings
White
Price
92,50 €
Log in for availability
1 * 500 ST
Product Details & Documents
Embedding rings made from ABS.
  • Dimensions: 41×28×11 mm
Available in the colours white, blue, yellow, green and pink.
Specifications
  • Colour:
    White