Orders LinkContactUsLinkComponent
Stopcocks for burettes, lateral
Stopcocks for burettes, lateral
1,5 mm, 12,5 NS
Catalog # LENZ02702501
Supplier:   Lenz Laborglas
Stopcocks for burettes, lateral
Catalog # LENZ02702501
Supplier:  Lenz Laborglas
Stopcocks for burettes, lateral
Stopcocks for burettes, lateral
1,5 mm, 12,5 NS
Price
41,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
NS stopcock made of borosilicate glass, facetted tip, with plug of solid glass or PTFE and screwthread retaining device.
  • Excellent chemical resistance
  • Excellent temperature resistance
Documents
Specifications
  • Hole Ø:
    1,5 mm
  • Spigot:
    12,5 NS