Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-channel pipettes, Multipet

Multi-channel pipettes, Multipet

50 - 300 µl, ±1,6 (50 µl); ±1,0 (300 µl) %, ≤ ±3,0 (50 µl); ±1,0 (300 µl) %
Catalog # KART1326.10
Supplier:   KARTELL
Multi-channel pipettes, Multipet
Catalog # KART1326.10
Supplier:  KARTELL
Multi-channel pipettes, Multipet
Multi-channel pipettes, Multipet

50 - 300 µl, ±1,6 (50 µl); ±1,0 (300 µl) %, ≤ ±3,0 (50 µl); ±1,0 (300 µl) %

Price
1 600,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These multi-channel micropipettes with air cushion are ideal for dispensing in multiwell plates for ELISA dosages, molecular biology and DNA amplifications.

  • Volume can be easily selected by turning the emptying button or the cylinder
  • Large display ensures easy reading of the set volume value
  • Cones are indipendent, so the required force for securing the 8 or 12 tips is uniformly distributed
  • Minimum pressure required for inserting and ejecting tips
  • Movement of the cones allows the micropipette to oscillate during the loading operations
  • Lower part can freely turn 360°, the ejector is easily adjustable to house a large variety of tips and can be removed for cleaning
  • Fully autoclavable at +121 °C for 20 minutes, without having to recalibrate them

Certifications: EC/IVD-compliant, in accordance with EN IS 8655

Delivery information: Supplied with individual calibration certificates.

Specifications
  • Volume:
    50 - 300 µl
  • Accuracy:
    ±1,6 (50 µl); ±1,0 (300 µl) %
  • Imprecision:
    ≤ ±3,0 (50 µl); ±1,0 (300 µl) %
Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Stand for 3 pipettors, 145×70 mm
    Stand for 3 pipettors, 145×70 mm
    KART1315
    Transparent
    Log in for availability
    1 * 1 ST - 95,70 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.