Orders LinkContactUsLinkComponent
Screw caps for screw thread tubes
Screw caps for screw thread tubes
GL 25
Catalog # LENZ01320025
Supplier:   Lenz Laborglas
Screw caps for screw thread tubes
Catalog # LENZ01320025
Supplier:  Lenz Laborglas
Screw caps for screw thread tubes
Screw caps for screw thread tubes
GL 25
Price
49,60 €
Log in for availability
1 * 10 ST
Product Details & Documents
PBT,red, with PTFE coated silicone sealing (VMQ).
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air
Specifications
  • Thread:
    GL 25
  • Seq:
    03