Attachable screw thread tubes
GL 14, 12 mm, 100 mm
LENZ01300014
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
DIN screw thread GL.
- Thread:GL 14
- Tube ext.Ø:12 mm
- Seq:01
- Total length:100 mm



