ContactUsLinkComponent
Blade, round deep-edged length: 25mm, width: 25mm, material thickness: 0.6mm2-sided ground-edge, 10 in magazine, double bevel edge-g round. 1 * 5 items
Catalog # MARO725.50
Supplier:  MARTOR ARGENTAX
Blade, round deep-edged length: 25mm, width: 25mm, material thickness: 0.6mm2-sided ground-edge, 10 in magazine, double bevel edge-g round. 1 * 5 items
Catalog # MARO725.50
Supplier:  MARTOR ARGENTAX
Frequently Bought Together