Orders LinkContactUsLinkComponent
Spray bottles
Spray bottles
1000 ml, Spray bottle, Transparent
Catalog # VITL95286
Supplier:   VITLAB
Spray bottles
Catalog # VITL95286
Supplier:  VITLAB
Spray bottles
Spray bottles
1000 ml, Spray bottle, Transparent
Price
16,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
White or translucent made from PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
Sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle, which can be regulated from the finest mist (nebulising) to a precise liquid jet. Range: approx. 3 to 4 metres. Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography.
Specifications
  • Capacity:
    1000 ml
  • Colour:
    Transparent
  • Description:
    Spray bottle
  • Seq:
    03