WHEATON® Rubber Stoppers
Stopper, natural red rubber, 13,0 mm, 20,0 mm
N° de catalogue 215-4714
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
Stoppers of various styles, sizes and rubber formulations.
- Manufactured from the highest quality raw materials
- Components manufactured to yield exceptional dimensional stability
- Autoclavable
Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.
Snap-on style provides maximum sealing for vials with a blow-back glass finish.
Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.
Documents
Spécifications
- Ø bottom:13,0 mm
- Ø top:20,0 mm
- Description:Stopper, natural red rubber
- Seq:17
- Cat. No.:215-4714
- Durometer hardness:45s



