Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(-)-Ribose ≥98%

Glass bottle for solids, 250 g
Catalog # A17894.30
D-(-)-Ribose ≥98%
Catalog # A17894.30
CAS Number:  50-69-1
D-(-)-Ribose ≥98%
D-(-)-Ribose ≥98%

Glass bottle for solids, 250 g

CAS Number:  50-69-1
Price
362,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 50-69-1 Molecular Formula: C5H10O5 Formula Weight: 150.13 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 82-86° Beilstein Reference: 1723081 Merck Reference: 14,8204 Optical Rotation: -20° (c=10 in water, 28h) MDL No.: MFCD00135453

  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:14,8204
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:82…86 °C
  • Density:0.8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-059-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.