Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nicotinamide ≥99%

Nicotinamide ≥99%

Glass bottle for solids, 1.000 g
Catalog # A15970.0B
Irritant
Nicotinamide ≥99%
Catalog # A15970.0B
CAS Number:  98-92-0
Nicotinamide ≥99%
Nicotinamide ≥99%

Glass bottle for solids, 1.000 g

CAS Number:  98-92-0
Price
190,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 98-92-0 Molecular Formula: C6H6N2O Formula Weight: 122.13 Flash Point: 182°(360°F) Physical Form: Powder Density: 1.40 Appearance: White Odour: Odourless Melting Point: 128-132° Beilstein Reference: 383619 Merck Reference: 14,6523 MDL No.: MFCD00006395

The amide of nicotinic acid (vitamin B-3)

Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006395
  • Molecular Weight:122.13 g/mol
  • Merck Index:14,6523
  • Boiling Pt:334 °C (1013 hPa)
  • Melting Pt:128…132 °C
  • Density:1.40
  • Flash Pt:182 °C (360 °F)
  • Storage Temperature:Ambient
  • EINECS:202-713-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.