Order Entry
Belgium
Orders LinkContactUsLinkComponent
Thymine 97%

Thymine 97%

Glass bottle for solids, 500 g
Catalog # A15879.36
Thymine 97%
Catalog # A15879.36
CAS Number:  65-71-4
Thymine 97%
Thymine 97%

Glass bottle for solids, 500 g

CAS Number:  65-71-4
Price
824,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 65-71-4 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 316-317° Beilstein Reference: 607626 Merck Reference: 14,9398 MDL No.: MFCD00006026

Synonyms:  5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
  • CAS Number:65-71-4
  • Formula:C₅H₆N₂O₂
  • MDL Number:MFCD00006026
  • Molecular Weight:126.11 g/mol
  • Merck Index:14,9398
  • Boiling Pt:389 °C (1013 hPa)
  • Melting Pt:316…317 °C
  • Density:1.454 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-616-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.