Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥99%

L(+)-Ascorbic acid ≥99%

Glass bottle for solids, 500 g
Catalog # A15613.36
L(+)-Ascorbic acid ≥99%
Catalog # A15613.36
CAS Number:  50-81-7
L(+)-Ascorbic acid ≥99%
L(+)-Ascorbic acid ≥99%

Glass bottle for solids, 500 g

CAS Number:  50-81-7
Price
102,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 190-193° dec. Beilstein Reference: 84272 Merck Reference: 14,830 Optical Rotation: +21° (c=10 in water) MDL No.: MFCD00064328

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:14,830
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…193 °C (dec.)
  • Density:1.65
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.