Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Chlorophenol ≥99%

4-Chlorophenol ≥99%

Glass bottle for solids, 100 g
Catalog # A15602.22
Irritant
Environment
4-Chlorophenol ≥99%
Catalog # A15602.22
CAS Number:  106-48-9
4-Chlorophenol ≥99%
4-Chlorophenol ≥99%

Glass bottle for solids, 100 g

CAS Number:  106-48-9
Price
42,20 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 106-48-9 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 115°(239°F) Odour: Phenol-like Boiling Point: 217-220° Beilstein Reference: 507004 Merck Reference: 14,2154 UN No.: 2020 MDL No.: MFCD00002318

Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Merck Index:14,2154
  • Boiling Pt:217…220 °C
  • Melting Pt:39…43 °C
  • Density:1.306
  • Flash Pt:115 °C (239 °F)
  • Storage Temperature:Ambient
  • EINECS:203-402-6
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.