Order Entry
Belgium
Orders LinkContactUsLinkComponent
2-Phenylethanol ≥98%

2-Phenylethanol ≥98%

Glass bottle for solids, 1.000 g
Catalog # A15241.0B
Toxic
Irritant
2-Phenylethanol ≥98%
Catalog # A15241.0B
CAS Number:  60-12-8
2-Phenylethanol ≥98%
2-Phenylethanol ≥98%

Glass bottle for solids, 1.000 g

CAS Number:  60-12-8
Price
77,20 €
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 98+% CAS No.: 60-12-8 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 101°(214°F) Physical Form: Liquid Density: 1.021 Refractive Index: 1.5330 Appearance: Colourless Melting Point: -27° Boiling Point: 219° Beilstein Reference: 1905732 Merck Reference: 14,7224 UN No.: 2810 MDL No.: MFCD00002886

Synonyms:  Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
  • CAS Number:60-12-8
  • Formula:C₆H₅CH₂CH₂OH
  • MDL Number:MFCD00002886
  • Molecular Weight:122.17 g/mol
  • Merck Index:14,7224
  • Boiling Pt:219 °C
  • Melting Pt:–27 °C
  • Density:1.021
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
  • EINECS:200-456-2
  • UN:2810
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications