Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cyclopentane 95%

Glass bottle, 2.500 mL
N° de catalogue A14697.0F
Flammable
Fournisseur:   Thermo Fisher Scientific
Cyclopentane 95%
N° de catalogue A14697.0F
Numéro CAS:  287-92-3
Cyclopentane 95%
Cyclopentane 95%

Glass bottle, 2.500 mL

Numéro CAS:  287-92-3
Prix
399,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 95% CAS No.: 287-92-3 Molecular Formula: C5H10 Formula Weight: 70.14 Flash Point: -37°(-34°F) Physical Form: Liquid Density: 0.748 Refractive Index: 1.4060 Appearance: Colourless Odour: Petrol-like Melting Point: -95° Boiling Point: 47-49° Beilstein Reference: 1900195 Merck Reference: 14,2741 UN No.: 1146 MDL No.: MFCD00001356

Synonyms:  Pentamethylene
  • CAS Number:287-92-3
  • Formula:C₅H₁₀
  • MDL Number:MFCD00001356
  • Molecular Weight:70.13 g/mol
  • Merck Index:14,2741
  • Boiling Pt:47…49 °C
  • Melting Pt:–95 °C
  • Density:0.748
  • Flash Pt:–37 °C (–34 °F)
  • Storage Temperature:Ambient
  • EINECS:206-016-6
  • UN:1146
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications