Acetanilide ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 103-84-4 Molecular Formula: C8H9NO Formula Weight: 135.17 Flash Point: 161°(321°F) Physical Form: Crystals Density: 1.121 Melting Point: 113-116° Boiling Point: 304-305° Beilstein Reference: 606468 Merck Reference: 14,50 MDL No.: MFCD00008674
Synonyms: N-Phenylacetamide
- CAS Number:103-84-4
- Formula:C₈H₉NO
- MDL Number:MFCD00008674
- Molecular Weight:135.17 g/mol
- Merck Index:14,50
- Boiling Pt:304…305 °C
- Melting Pt:113…116 °C
- Density:1.121
- Flash Pt:161 °C (321 °F)
- Storage Temperature:Ambient
- EINECS:203-150-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



