Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Biotin ≥98%

D-(+)-Biotin ≥98%

Glass bottle for solids, 1 g
Catalog # A14207.03
D-(+)-Biotin ≥98%
Catalog # A14207.03
CAS Number:  58-85-5
D-(+)-Biotin ≥98%
D-(+)-Biotin ≥98%

Glass bottle for solids, 1 g

CAS Number:  58-85-5
Price
85,60 €
Connexion Pour la disponibilité
1 * 1 g
Product Details & Documents

Purity: 98+% CAS No.: 58-85-5 Molecular Formula: C10H16N2O3S Formula Weight: 244.32 Storage Temperature: 2°C to 8°C Melting Point: 224-226° Beilstein Reference: 86838 Merck Reference: 14,1231 Optical Rotation: +91° (c=1 in 0.1N NaOH) MDL No.: MFCD00005541

Essential vitamin that is important for amino acid and energy metabolism

Synonymes:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • Numéro CAS:58-85-5
  • Formule:C₁₀H₁₆N₂O₃S
  • Numéro MDL:MFCD00005541
  • Poids moléculaire:244.31 g/mol
  • Index Merck:14,1231
  • Point de fusion:224…226 °C
  • Densité:1.41 g/cm³ (20 °C)
  • Température de stockage:Refrigerator
  • EINECS:200-399-3
Spécifications

Spécifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.