Order Entry
Belgium
Orders LinkContactUsLinkComponent
Creatinine ≥98%

Creatinine ≥98%

Glass bottle for solids, 250 g
Catalog # B23097.30
Irritant
Creatinine ≥98%
Catalog # B23097.30
CAS Number:  60-27-5
Creatinine ≥98%
Creatinine ≥98%

Glass bottle for solids, 250 g

CAS Number:  60-27-5
Price
394,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 60-27-5 Molecular Formula: C4H7N3O Formula Weight: 113.12 Odour: Odourless pH: 7.5-8.5 Melting Point: ca 255° dec. Beilstein Reference: 112061 Merck Reference: 14,2569 MDL No.: MFCD00059730

Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Merck Index:14,2569
  • Melting Pt:∼ 255 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-466-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications