Creatinine ≥98%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 60-27-5 Molecular Formula: C4H7N3O Formula Weight: 113.12 Odour: Odourless pH: 7.5-8.5 Melting Point: ca 255° dec. Beilstein Reference: 112061 Merck Reference: 14,2569 MDL No.: MFCD00059730
Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
- CAS Number:60-27-5
- Formula:C₄H₇N₃O
- MDL Number:MFCD00059730
- Molecular Weight:113.12 g/mol
- Merck Index:14,2569
- Melting Pt:∼ 255 °C (dec.)
- Storage Temperature:Ambient
- EINECS:200-466-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



