Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Chlorophenol red

Glass bottle for solids, 5 g
N° de catalogue B21623.06
Irritant
Fournisseur:   Thermo Fisher Scientific
Chlorophenol red
N° de catalogue B21623.06
Numéro CAS:  4430-20-0
Chlorophenol red
Chlorophenol red

Glass bottle for solids, 5 g

Numéro CAS:  4430-20-0
Prix
46,80 €
Log in for availability
1 * 5 g
Product Details & Documents

CAS No.: 4430-20-0 Molecular Formula: C19H12Cl2O5S Formula Weight: 432.28 Beilstein Reference: 354053 MDL No.: MFCD00005877

Synonyms:  3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
  • CAS Number:4430-20-0
  • Formula:C₁₉H₁₂Cl₂O₅S
  • MDL Number:MFCD00005877
  • Molecular Weight:423.27 g/mol
  • Melting Pt:∼261 °C
  • Storage Temperature:Ambient
  • EINECS:224-619-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications