Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Aspartic acid ≥99%

D-(-)-Aspartic acid ≥99%

Glass bottle for solids, 25 g
Catalog # B21184.14
D-(-)-Aspartic acid ≥99%
Catalog # B21184.14
CAS Number:  1783-96-6
D-(-)-Aspartic acid ≥99%
D-(-)-Aspartic acid ≥99%

Glass bottle for solids, 25 g

CAS Number:  1783-96-6
Price
101,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 1783-96-6 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Melting Point: 300° dec. Beilstein Reference: 1723529 Merck Reference: 14,840 Optical Rotation: -25° (c=1 in 1N HCl) MDL No.: MFCD00063081

Synonyms:  D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
  • CAS Number:1783-96-6
  • Formula:C₄H₇NO₄
  • MDL Number:MFCD00063081
  • Molecular Weight:133.1 g/mol
  • Merck Index:14,840
  • Melting Pt:>300 °C (dec.)
  • Density:1.66
  • Storage Temperature:Ambient
  • EINECS:217-234-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.