Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(+)-Galactose ≥98%

Glass bottle for solids, 250 g
Catalog # A12813.30
D-(+)-Galactose ≥98%
Catalog # A12813.30
CAS Number:  59-23-4
D-(+)-Galactose ≥98%
D-(+)-Galactose ≥98%

Glass bottle for solids, 250 g

CAS Number:  59-23-4
Price
129,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98% CAS No.: 59-23-4 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 164-168° Beilstein Reference: 1724619 Merck Reference: 14,4335 Optical Rotation: +80° (c=5 in water, 24h) MDL No.: MFCD00151230

Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,4335
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:164…168 °C
  • Density:1.50
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications