Sodium nitrate ≥98%
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Odour: Odourless Melting Point: ca 308° UN No.: 1498 MDL No.: MFCD00011119
- CAS Number:7631-99-4
- Formula:NaNO₃
- MDL Number:MFCD00011119
- Molecular Weight:84.99 g/mol
- Merck Index:14,8647
- Boiling Pt:380 °C (1013 hPa)
- Melting Pt:∼308 °C
- Density:2.26
- Storage Temperature:Ambient
- EINECS:231-554-3
- UN:1498
- ADR:5.1,III
- REACH:01-2119488221-41
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g



