Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

2-Deoxy-D(-)-ribose ≥99%

Glass bottle for solids, 1 g
Catalog # A11990.03
2-Deoxy-D(-)-ribose ≥99%
Catalog # A11990.03
CAS Number:  533-67-5
2-Deoxy-D(-)-ribose ≥99%
2-Deoxy-D(-)-ribose ≥99%

Glass bottle for solids, 1 g

CAS Number:  533-67-5
Price
29,60 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 99% CAS No.: 533-67-5 Molecular Formula: C5H10O4 Formula Weight: 134.13 Melting Point: 76-85° Beilstein Reference: 1721978 Merck Reference: 14,2908 Optical Rotation: -56° (c=1 in water, 24h) MDL No.: MFCD00135904

Synonyms:  2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
  • CAS Number:533-67-5
  • Formula:C₅H₁₀O₄
  • MDL Number:MFCD00135904
  • Molecular Weight:134.13 g/mol
  • Merck Index:14,2908
  • Boiling Pt:292 °C (1013 hPa)
  • Melting Pt:76…85 °C
  • Storage Temperature:Ambient
  • EINECS:208-573-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications