Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Fluorophenol ≥99%

4-Fluorophenol ≥99%

Glass bottle for solids, 500 g
Catalog # A11945.36
Corrosive
Irritant
4-Fluorophenol ≥99%
Catalog # A11945.36
CAS Number:  371-41-5
4-Fluorophenol ≥99%
4-Fluorophenol ≥99%

Glass bottle for solids, 500 g

CAS Number:  371-41-5
Price
499,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 371-41-5 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 68°(154°F) Odour: Phenol-like Boiling Point: 185-188° Beilstein Reference: 1362752 UN No.: 2923 MDL No.: MFCD00002316

Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185…188 °C
  • Melting Pt:44…50 °C
  • Density:1.22
  • Flash Pt:68 °C (154 °F)
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.