Order Entry
Belgium
Orders LinkContactUsLinkComponent
Quinoline ≥97%

Quinoline ≥97%

Glass bottle for solids, 100 g
Catalog # A11545.22
Toxic
Health
Irritant
Quinoline ≥97%
Catalog # A11545.22
CAS Number:  91-22-5
Quinoline ≥97%
Quinoline ≥97%

Glass bottle for solids, 100 g

CAS Number:  91-22-5
Price
57,30 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Refractive Index: 1.6260 Melting Point: -15° Boiling Point: 237° Solubility: More soluble in hot than cold water. Miscible with alcohol, ether Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736

  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:14,8068
  • Boiling Pt:237 °C
  • Melting Pt:–15 °C
  • Density:1.090
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.