Mesitylene ≥98%
Glass bottle, 2.500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 108-67-8 Molecular Formula: C9H12 Formula Weight: 120.20 Flash Point: 44°(111°F) Refractive Index: 1.4990 Odour: Unpleasant Melting Point: -45° Boiling Point: 163-165° Beilstein Reference: 906806 Merck Reference: 14,5907 UN No.: 2325 MDL No.: MFCD00008538
Synonyms: 1,3,5-Trimethylbenzene
- CAS Number:108-67-8
- Formula:C₉H₁₂
- MDL Number:MFCD00008538
- Molecular Weight:120.19 g/mol
- Merck Index:14,5907
- Boiling Pt:163…165 °C
- Melting Pt:–45 °C
- Density:0.865
- Flash Pt:44 °C (111 °F)
- Storage Temperature:Ambient
- EINECS:203-604-4
- UN:2325
- ADR:3,III
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2.500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



