Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin ≥99%

Vanillin ≥99%

Glass bottle for solids, 500 g
Catalog # A11169.36
Irritant
Vanillin ≥99%
Catalog # A11169.36
CAS Number:  121-33-5
Vanillin ≥99%
Vanillin ≥99%

Glass bottle for solids, 500 g

CAS Number:  121-33-5
Price
130,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 121-33-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 153°(307°F) Physical Form: Crystalline powder Density: 1.06 Sensitivity: Air and Light Sensitive Appearance: White Melting Point: 81-83° Boiling Point: 170°/15mm Beilstein Reference: 472792 Merck Reference: 14,9932 MDL No.: MFCD00006942

Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,9932
  • Boiling Pt:170 °C (15 mmHg)
  • Melting Pt:81…83 °C
  • Density:1.06
  • Flash Pt:153 °C (307 °F)
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.