Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin ≥99%

Vanillin ≥99%

Glass bottle for solids, 100 g
Catalog # A11169.22
Irritant
Vanillin ≥99%
Catalog # A11169.22
CAS Number:  121-33-5
Vanillin ≥99%
Vanillin ≥99%

Glass bottle for solids, 100 g

CAS Number:  121-33-5
Price
40,70 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 121-33-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 153°(307°F) Physical Form: Crystalline powder Density: 1.06 Sensitivity: Air and Light Sensitive Appearance: White Melting Point: 81-83° Boiling Point: 170°/15mm Beilstein Reference: 472792 Merck Reference: 14,9932 MDL No.: MFCD00006942

Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,9932
  • Boiling Pt:170 °C (15 mmHg)
  • Melting Pt:81…83 °C
  • Density:1.06
  • Flash Pt:153 °C (307 °F)
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications