Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Mannose ≥99%

D-(+)-Mannose ≥99%

Glass bottle for solids, 500 g
Catalog # A10842.36
D-(+)-Mannose ≥99%
Catalog # A10842.36
CAS Number:  3458-28-4
D-(+)-Mannose ≥99%
D-(+)-Mannose ≥99%

Glass bottle for solids, 500 g

CAS Number:  3458-28-4
Price
642,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 3458-28-4 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 127-133° Solubility: Soluble in water Beilstein Reference: 1564373 Merck Reference: 14,5747 Optical Rotation: +13.8° (c=10 in water, 24h) MDL No.: MFCD00064122

Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,5747
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:127…133 °C
  • Density:1.54
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.