Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Mannose ≥99%

D-(+)-Mannose ≥99%

Glass bottle for solids, 100 g
Catalog # A10842.22
D-(+)-Mannose ≥99%
Catalog # A10842.22
CAS Number:  3458-28-4
D-(+)-Mannose ≥99%
D-(+)-Mannose ≥99%

Glass bottle for solids, 100 g

CAS Number:  3458-28-4
Price
152,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 3458-28-4 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 127-133° Solubility: Soluble in water Beilstein Reference: 1564373 Merck Reference: 14,5747 Optical Rotation: +13.8° (c=10 in water, 24h) MDL No.: MFCD00064122

Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,5747
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:127…133 °C
  • Density:1.54
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications