Order Entry
Belgium
Orders LinkContactUsLinkComponent
Caffeine ≥99%

Caffeine ≥99%

Glass bottle for solids, 100 g
Catalog # A10431.22
Irritant
Caffeine ≥99%
Catalog # A10431.22
CAS Number:  58-08-2
Caffeine ≥99%
Caffeine ≥99%

Glass bottle for solids, 100 g

CAS Number:  58-08-2
Price
34,40 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Solubility: Soluble in pyrrole, THF-H2O mixture, ethyl acetate Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758

Has inhibitory action against lung and colon tumorigenses, and virus induction

Synonyms:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS Number:58-08-2
  • Formula:C₈H₁₀N₄O₂
  • MDL Number:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck Index:14,1636
  • Melting Pt:234…236 °C
  • Density:1.230
  • Storage Temperature:Ambient
  • EINECS:200-362-1
  • UN:1544
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.