Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(-)-Proline ≥99%

L(-)-Proline ≥99%

Glass bottle for solids, 25 g
Catalog # A10199.14
L(-)-Proline ≥99%
Catalog # A10199.14
CAS Number:  147-85-3
L(-)-Proline ≥99%
L(-)-Proline ≥99%

Glass bottle for solids, 25 g

CAS Number:  147-85-3
Price
41,80 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 147-85-3 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: ca 225° dec. Beilstein Reference: 80810 Merck Reference: 14,7780 Optical Rotation: -84° (c=1 in water) MDL No.: MFCD00064318

Synonyms:  (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
  • CAS Number:147-85-3
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00064318
  • Molecular Weight:115.13 g/mol
  • Merck Index:14,7780
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:∼ 225 °C (dec.)
  • Density:1.36
  • Storage Temperature:Ambient
  • EINECS:205-702-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications