Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 28%

Ammonia solution 28%

Glass bottle, 2.500 mL
Catalog # L13168.0F
Corrosive
Irritant
Environment
Ammonia solution 28%
Catalog # L13168.0F
CAS Number:  1336-21-6
Ammonia solution 28%
Ammonia solution 28%

Glass bottle, 2.500 mL

CAS Number:  1336-21-6
Price
64,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Odour: Ammonia-like pH: 12 Merck Reference: 14,494 MDL No.: MFCD00011418

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Merck Index:14,494
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.90
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications