Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer HF-Improved

Buffer HF-Improved, 250 mL
Catalog # 44627.AK
Buffer HF-Improved
Catalog # 44627.AK
CAS Number:  
Buffer HF-Improved
Buffer HF-Improved

Buffer HF-Improved, 250 mL

CAS Number:  
Price
41,40 €
Connexion Pour la disponibilité
1 * 250 mL
Product Details & Documents

Buffer used for pretreatment of planar silicon devices when plating with nickel plating solution products 44069 and 44070.

Density: 1.12
Appearance: Colorless
Odor: Acrid
Boiling Point: 103…105°
UN No.: 2817

Spécifications

Spécifications

  • Pk:
    250 mL
  • Description:
    Buffer HF-Improved
  • UN No.:
    2817
  • Cat. No.:
    44627.AK
  • Odour/scent:
    Acrid
  • MDL No.:
    MFCD00132734
  • Density:
    1.12 g/mL
  • Appearance:
    Colourless

Spécifications