Calibration software, EASYCAL™ 5.0
The EASYCAL™ 5.0 calibration software facilitates the monitoring of Liquid Handling instruments (like piston-stroke devices such as pipettes, dispensers, burettes, and manual dispensers), as well as volumetric instruments made of glass or plastic, based on GLP/GMP and DIN EN ISO 9001. EASYCAL™ 5 software can be used for BRAND instruments as well as instruments from other manufacturers.
- Simple: Can be used with pipettes and instruments of all manufacturers
- Easy: Automatic calculation using the up-to-date standards
- Efficient: Professional and transparent recording of all parameters
EASYCAL™ 5 automatically calculates and compares all values against the tolerances from current standards or individually-defined limits. The tolerances of numerous devices and the interface settings of over 100 test devices (e.g. balances) are already stored for you. Choose between a stand-alone license for working on one workstation or a client/server version for parallel, distributed work on multiple workstations.
The stand-alone license is recommended for small labs where calibration is completed by one person. The client/server version and its floating licenses are an excellent solution for larger labs where calibration will be carried out at multiple workstations.
Functions: Testing of liquid handling devices and volumetric instruments made of glass and plastic according to standards such as ISO 8655, ISO 4787, and others. Open software, suitable for all volumetric instruments - from any manufacturer. Extensive library with device specifications from many well-known manufacturers - can be expanded and modified by the user. Flexible test plans for your needs.
Extensive library of test plans included to help you get started with EASYCAL™ 5 and minimise data entry time. Device management - quickly and easily search and find the owner, test history, and next test date. Continuous control of the current actual state during testing by graphical representations and real time calculation of statistical values. Reminder function for upcoming tests with automatic email notifications sent to device owner. Integration of the address data of your customers and suppliers in a business partner database. User management with user roles (e.g., inspector, supervisor, and system administrator) and access restriction to EASYCAL functions. Dual-control principle for the release of critical data such as test plans, calibration orders before certificate printing, and device specification. RS232 interface connection for measuring devices such as scales, thermometers, barometers, and hygrometers with automatic data transmission of the measured values. Customise certificates and audit trails to your needs in the integrated certificate editor.
- Description:EASYCAL™ 5.0, stand-alone licence, includes lifetime updates
- Seq:01
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.