Mailing container, with screw cap, square
Mailing container, square, 117×35 mm, Ø opening: 28 mm, with cap, Natural
SARS78.572
Marketsource
: SARSTEDT
Containers for safe postage and transportation of clinical samples (blood, urine, swabs and faeces tubes).
- Tight screw caps ensures the safety
- Made of neutral HDPE
- One side of each container can be printed
- Available with or without absorbent liners
Suitable for 30 ml liquid sample and faeces tubes of 107ר25 mm.
- Colour:Natural
- Description:Mailing container, square, 117×35 mm, Ø opening: 28 mm, with cap
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



