Order Entry
Belgium
Orders LinkContactUsLinkComponent
Urine sample container

Urine sample container

Container, brown, with black cap mm, 500 ml, 132 mm
Catalog # SARS77.582
Marketsource
Supplier:   SARSTEDT
Urine sample container
Catalog # SARS77.582
Supplier:  SARSTEDT
Marketsource
Urine sample container

Container, brown, with black cap mm, 500 ml, 132 mm

Price
Inquire For Price
Log in for availability
1 * 100 ST
Product Details & Documents

Container made of PE, opaque, white with black HDPE screw cap.

  • Good chemical resistance
  • Good temperature resistance

For collection and storage of urine.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Capacity:
    500 ml
  • Description:
    Container, brown, with black cap mm
  • Height:
    132 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.