Orders LinkContactUsLinkComponent
Microtubes, with Thread
Micro tubes with neutral screw cap enclosed, without skirted base, 0,5 ml, 5000/case
N° de catalogue SARS72.733.001
Marketsource
Fournisseur:   SARSTEDT
Microtubes, with Thread
N° de catalogue SARS72.733.001
Fournisseur:  SARSTEDT
Marketsource
Microtubes, with Thread
Micro tubes with neutral screw cap enclosed, without skirted base, 0,5 ml, 5000/case
Prix
410,26 €
Log in for availability
1 * 5.000 ST
Product Details & Documents
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.
Specifications
  • Packed:
    5000/case
  • Capacity:
    0,5 ml
  • Description:
    Micro tubes with neutral screw cap enclosed, without skirted base
  • Seq:
    8